Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008574.3 | NP_032600.2 | 2 | 210 | ||
GenBank mRNA | AK077172.1 | - | 2 | 181 | ||
BC055104.1 | - | 2 | 180 | 6773089 | ||
BC061167.1 | - | 2 | 160 | 6771333 | ||
BU936528.1 | - | 2 | 213 | |||
M29603.1 | - | 2 | 183 | |||
S49659.1 | - | 2 | 180 | |||
S49660.1 | - | 330 |