Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252437.1 | NP_001239366.1 | 3, 4 | 584 | ||
NM_029657.4 | NP_083933.1 | 3, 4 | 581 | |||
GenBank mRNA | AK011747.2 | - | 3, 4 | 374 | ||
AK088533.1 | - | 3, 4 | 364 | |||
AK150176.1 | - | 4, 5 | 417 | |||
AK153407.1 | - | 3, 4 | 405 | |||
BC046830.1 | - | 3, 4 | 430 | 6811606 |