Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meis1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193271.1 | NP_001180200.1 | 6 | 1319 | ||
NM_010789.3 | NP_034919.1 | 6 | 1319 | |||
GenBank mRNA | AB221660.1 | - | 5 | 582 | ||
AK032947.1 | - | 5 | 1130 | |||
AK049720.1 | - | 5 | 1131 | |||
AK132298.1 | - | 6 | 1099 | |||
AK134321.1 | - | 6 | 1314 | |||
AK140748.1 | - | 6 | 1323 | |||
AK146680.1 | - | 2 | 127 | |||
AK168684.1 | - | 6 | 1311 | |||
BC023689.1 | - | 6 | 1315 | 5344307 |