Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Melk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010790.2 | NP_034920.2 | 14 | 1509 | ||
GenBank mRNA | AK010498.1 | - | 14 | 1475 | ||
AK011932.1 | - | 14 | 1509 | |||
AK129076.1 | - | 14 | 1462 | |||
AK137000.1 | - | 14 | 1535 | |||
AK145021.1 | - | 14 | 1444 | |||
AK145316.1 | - | 14 | 1418 | |||
AK161029.1 | - | 14 | 1518 | |||
AK164138.1 | - | 14 | 1476 | |||
AK167457.1 | - | 14 | 1433 | |||
BC020416.1 | - | 13 | 1344 | |||
BC085276.1 | - | 14 | 1542 | 30443236 | ||
L76158.1 | - | 14 | 1440 | |||
X95351.1 | - | 14 | 1447 |