Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Melk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010790.2 | NP_034920.2 | 5 | 532 | ||
GenBank mRNA | AK010498.1 | - | 5 | 499 | ||
AK011932.1 | - | 5 | 532 | |||
AK129076.1 | - | 5 | 485 | |||
AK137000.1 | - | 5 | 557 | |||
AK145021.1 | - | 5 | 467 | |||
AK145316.1 | - | 5 | 441 | |||
AK161029.1 | - | 5 | 541 | |||
AK164138.1 | - | 5 | 499 | |||
AK167457.1 | - | 5 | 456 | |||
BC020416.1 | - | 4 | 367 | |||
BC085276.1 | - | 5 | 565 | 30443236 | ||
L76158.1 | - | 5 | 463 | |||
X95351.1 | - | 5 | 470 |