Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110253.2 | NP_001103723.2 | 5 | 533 | ||
NM_148925.3 | NP_683727.3 | 6 | 780 | |||
GenBank mRNA | AJ428065.1 | - | 6 | 685 | ||
AK081259.1 | - | 5 | 516 | |||
BC053712.1 | - | 4 | 441 |