Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008584.3 | NP_032610.1 | 2 | 951 | ||
GenBank mRNA | AK028352.1 | - | 2 | 952 | ||
AK031152.1 | - | 2 | 908 | |||
AK142690.1 | - | 2 | 830 | |||
BC002076.1 | - | 2 | 850 | 3591924 | ||
Z16406.1 | - | 2 | 829 |