Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mertk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008587.1 | NP_032613.1 | 19 | 2789 | ||
GenBank mRNA | AK029009.1 | - | 8 | 991 | ||
AK030158.1 | - | 11 | 1488 | |||
AK079252.1 | - | 2 | 402 | |||
L11625.1 | - | 10 | 1327 | |||
U21301.1 | - | 19 | 2789 |