Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005347.4 | NP_005338.1 | 6 | 1438 | ||
GenBank mRNA | AF188611.1 | - | 6 | 1123 | ||
AF216292.1 | - | 6 | 1177 | |||
AJ271729.1 | - | 6 | 1190 | |||
BC020235.1 | - | 6 | 1381 | 5020098 | ||
EU794617.1 | - | 6 | 1190 | |||
X87949.1 | - | 6 | 1396 |