Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfge8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045489.1 | NP_001038954.1 | 6 | 793 | ||
NM_008594.2 | NP_032620.2 | 7 | 904 | |||
GenBank mRNA | AB021130.2 | - | 7 | 811 | ||
AB025280.1 | - | 6 | 700 | |||
AK087555.1 | - | 6 | 783 | |||
AK089211.1 | - | 6 | 759 | |||
AK145183.1 | - | 6 | 757 | |||
AK148371.1 | - | 6 | 782 | |||
AK152088.1 | - | 7 | 893 | |||
AK155047.1 | - | 7 | 902 | |||
AK169997.1 | - | 6 | 757 | |||
AK170935.1 | - | 6 | 757 | |||
AK171143.1 | - | 7 | 868 | |||
AK189618.1 | - | 2 | 178 | |||
AK207561.1 | - | 1 | 50 | |||
AK212128.1 | - | 1 | 50 | |||
AK212659.1 | - | 1 | 50 | |||
AK213407.1 | - | 1 | 50 | |||
AK214404.1 | - | 1 | 50 | |||
AK219710.1 | - | 1 | 50 | |||
BC003892.1 | - | 6 | 737 | 3593310 | ||
BC003904.1 | - | 6 | 732 | 3601565 | ||
BC018577.1 | - | 6 | 730 | |||
BC037475.1 | - | 3 | 1119 | |||
M38337.1 | - | 7 | 856 | |||
Y11684.2 | - | 6 | 700 |