Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010795.4 | NP_034925.2 | 2 | 247 | ||
GenBank mRNA | AK081850.1 | - | 2 | 311 | ||
BC053040.1 | - | 2 | 157 | 6406273 | ||
BC059828.1 | - | 1 | 139 | 5706186 | ||
CJ174488.1 | - | 2 | 247 |