Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204252.1 | NP_001191181.1 | 7, 8 | 821 | ||
NM_010796.3 | NP_034926.1 | 7, 8 | 818 | |||
GenBank mRNA | AK140762.1 | - | 6, 7 | 1791 | ||
AK170779.1 | - | 7, 8 | 778 | |||
BC014811.1 | - | 7, 8 | 651 | 4456238 | ||
S36676.1 | - | 7, 8 | 667 |