Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008598.2 | NP_032624.1 | 3, 4 | 379 | ||
GenBank mRNA | AB092489.1 | - | 2, 3 | 290 | ||
AB092490.1 | - | 2, 3 | 290 | |||
AK090389.1 | - | 369 | ||||
BC031888.1 | - | 3, 4 | 348 | 5372377 | ||
BC096557.1 | - | 3, 4 | 382 | 1366095 | ||
M84524.1 | - | 2, 3 | 305 |