Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aff1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080798.1 | NP_001074267.1 | 11 | 2050 | ||
NM_133919.3 | NP_598680.3 | 11 | 2061 | |||
GenBank mRNA | AF013131.1 | - | 10 | 1876 | ||
AF074266.1 | - | 11 | 1891 | |||
AK139900.1 | - | 3 | 737 | |||
AK143742.1 | - | 11 | 2050 | |||
AY749168.1 | - | 11 | 1894 | |||
BC131665.1 | - | 11 | 1894 | 40110795 |