Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008605.3 | NP_032631.3 | 10 | 1430 | ||
GenBank mRNA | AK089309.1 | - | 10 | 1399 | ||
AK089452.1 | - | 10 | 1399 | |||
AK089509.1 | - | 11 | 1273 | |||
AK089523.1 | - | 10 | 1399 | |||
AK090051.1 | - | 10 | 1401 | |||
AK150596.1 | - | 10 | 1399 | |||
AK151021.1 | - | 10 | 1399 | |||
AK151273.1 | - | 10 | 1399 | |||
AK151933.1 | - | 10 | 1399 | |||
AK155139.1 | - | 10 | 1398 | |||
AK155743.1 | - | 10 | 1399 | |||
AK155747.1 | - | 10 | 1399 | |||
AK155748.1 | - | 10 | 1399 | |||
AK155841.1 | - | 10 | 1457 | |||
AK155865.1 | - | 10 | 1398 | |||
AK170360.1 | - | 10 | 1399 | |||
AK170364.1 | - | 10 | 1399 | |||
AK170500.1 | - | 11 | 1273 | |||
AK170798.1 | - | 10 | 1398 | |||
AK170804.1 | - | 10 | 1398 | |||
AK170976.1 | - | 10 | 1399 | |||
AK171006.1 | - | 10 | 1399 | |||
AK171032.1 | - | 10 | 1398 | |||
AK171036.1 | - | 10 | 1399 | |||
AK171176.1 | - | 10 | 1399 | |||
AK171428.1 | - | 10 | 1399 | |||
AK171435.1 | - | 9 | 2446 | |||
AK171447.1 | - | 10 | 1399 | |||
AK171448.1 | - | 10 | 1399 | |||
AK171449.1 | - | 10 | 1399 | |||
AK171450.1 | - | 10 | 1399 | |||
AK171466.1 | - | 10 | 1399 | |||
AK171468.1 | - | 10 | 1399 | |||
AK171470.1 | - | 10 | 1399 | |||
AK171472.1 | - | 10 | 1399 | |||
AK171485.1 | - | 10 | 1399 | |||
AK171518.1 | - | 10 | 1399 | |||
AK171542.1 | - | 10 | 1399 | |||
AK171570.1 | - | 10 | 1399 | |||
BC019135.1 | - | 10 | 1393 | 5037674 | ||
CT010377.1 | - | 10 | 1346 | |||
M82831.1 | - | 10 | 1346 |