Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001540.3 | NP_001531.1 | 1 | 450 | ||
GenBank mRNA | AB020027.1 | - | 1 | 320 | ||
AF086135.1 | - | 1 | 363 | |||
AK296890.1 | - | 2 | 230 | |||
AK311894.1 | - | 1 | 333 | |||
AL050380.1 | - | 2 | 181 | |||
BC000510.2 | - | 1 | 394 | 2822325 | ||
BC012292.1 | - | 1 | 368 | 3862626 | ||
BC012768.2 | - | 1 | 317 | 3689220 | ||
BC014920.1 | - | 1 | 254 | |||
BC073768.1 | - | 1 | 315 | 5444848 | ||
BP231480.1 | - | 1 | 450 | |||
BT019888.1 | - | 1 | 295 | |||
CR407614.1 | - | 1 | 295 | |||
CR536489.1 | - | 1 | 295 | |||
FJ224323.1 | - | 1 | 402 | |||
U90906.1 | - | 1 | 402 | |||
X54079.1 | - | 1 | 347 |