Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008608.3 | NP_032634.3 | 6 | 1221 | ||
GenBank mRNA | AK040968.1 | - | 1194 | |||
AK088476.1 | - | 6 | 1221 | |||
AK134340.1 | - | 6 | 1152 | |||
AK138611.1 | - | 6 | 1162 | |||
AK149636.1 | - | 6 | 1150 | |||
AK149907.1 | - | 6 | 1201 | |||
AK149988.1 | - | 6 | 1121 | |||
AK155117.1 | - | 6 | 1223 | |||
AK163601.1 | - | 6 | 1213 | |||
AK165014.1 | - | 6 | 1222 | |||
AK212086.1 | - | 2 | 172 | |||
BC076638.1 | - | 6 | 1213 | 30658833 | ||
DQ249870.1 | - | 6 | 959 | |||
U54984.1 | - | 6 | 1205 | |||
X83536.2 | - | 6 | 1131 |