Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008608.3 | NP_032634.3 | 5 | 1070 | ||
GenBank mRNA | AK040968.1 | - | 1043 | |||
AK088476.1 | - | 5 | 1070 | |||
AK134340.1 | - | 5 | 1001 | |||
AK138611.1 | - | 5 | 1011 | |||
AK149636.1 | - | 5 | 999 | |||
AK149907.1 | - | 5 | 1050 | |||
AK149988.1 | - | 5 | 970 | |||
AK155117.1 | - | 5 | 1072 | |||
AK163601.1 | - | 5 | 1062 | |||
AK165014.1 | - | 5 | 1071 | |||
AK180818.1 | - | 1 | 47 | |||
AK182471.1 | - | 1 | 46 | |||
AK212086.1 | - | 1 | 21 | |||
BC076638.1 | - | 5 | 1062 | 30658833 | ||
DQ249870.1 | - | 5 | 808 | |||
U54984.1 | - | 5 | 1054 | |||
X83536.2 | - | 5 | 980 |