Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019724.3 | NP_062698.2 | 6 | 1291 | ||
GenBank mRNA | AB021228.1 | - | 6 | 1106 | ||
AF282844.1 | - | 6 | 1058 | |||
AK048543.1 | - | 6 | 1171 | |||
AK052039.1 | - | 1 | 2663 | |||
AK053598.1 | - | 5 | 3666 | |||
AK077939.1 | - | 1334 | ||||
BC057926.1 | - | 6 | 1293 | 5292478 |