Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019724.3 | NP_062698.2 | 4 | 658 | ||
GenBank mRNA | AB021228.1 | - | 4 | 473 | ||
AF282844.1 | - | 4 | 425 | |||
AK046170.1 | - | 4 | 517 | |||
AK048543.1 | - | 4 | 538 | |||
AK053598.1 | - | 4 | 548 | |||
AK077939.1 | - | 702 | ||||
BC057926.1 | - | 4 | 660 | 5292478 |