Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008611.4 | NP_032637.3 | 9 | 1359 | ||
GenBank mRNA | AK089234.1 | - | 9 | 1330 | ||
AK089336.1 | - | 9 | 1334 | |||
AK089381.1 | - | 9 | 1331 | |||
AK089516.1 | - | 9 | 1320 | |||
AK137468.1 | - | 9 | 1331 | |||
AK154937.1 | - | 9 | 1330 | |||
AK155740.1 | - | 9 | 1331 | |||
AK155832.1 | - | 9 | 1330 | |||
AK155901.1 | - | 9 | 1334 | |||
AK170177.1 | - | 9 | 1330 | |||
AK170388.1 | - | 9 | 1332 | |||
AK170760.1 | - | 9 | 1330 | |||
AK171168.1 | - | 9 | 1330 | |||
AK171613.1 | - | 9 | 1332 | |||
BC042742.1 | - | 9 | 1340 | 5032770 | ||
U96696.1 | - | 9 | 1325 | |||
Y13342.1 | - | 9 | 1257 |