Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008613.3 | NP_032639.1 | 9 | 1476 | ||
GenBank mRNA | AK013048.1 | - | 9 | 1429 | ||
AK132772.1 | - | 9 | 1451 | |||
BC046624.1 | - | 9 | 1389 | 6314904 | ||
D14849.1 | - | 9 | 1507 |