Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008618.3 | NP_032644.3 | 8 | 1013 | ||
GenBank mRNA | AK002395.1 | - | 8 | 887 | ||
AK002404.1 | - | 8 | 871 | |||
AK005237.1 | - | 8 | 908 | |||
AK008270.1 | - | 8 | 885 | |||
AK164785.1 | - | 8 | 894 | |||
AK168545.1 | - | 8 | 851 | |||
BC050940.1 | - | 8 | 879 | 6820601 | ||
M29462.1 | - | 8 | 892 |