Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008618.3 | NP_032644.3 | 2 | 259 | ||
GenBank mRNA | AK002395.1 | - | 2 | 138 | ||
AK002404.1 | - | 2 | 118 | |||
AK005237.1 | - | 2 | 154 | |||
AK008270.1 | - | 2 | 131 | |||
AK164785.1 | - | 2 | 140 | |||
AK168545.1 | - | 2 | 97 | |||
BC050940.1 | - | 2 | 125 | 6820601 | ||
M29462.1 | - | 2 | 138 |