Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000034.3 | NP_000025.1 | 14 | 2323 | ||
NM_001127617.2 | NP_001121089.1 | 9 | 1545 | |||
NM_001243177.1 | NP_001230106.1 | 10 | 1666 | |||
NM_184041.2 | NP_908930.1 | 9 | 1512 | |||
NM_184043.2 | NP_908932.1 | 9 | 1484 | |||
XM_011545768.2 | XP_011544070.1 | 11 | 2186 | |||
GenBank mRNA | AK026577.1 | - | 9 | 1361 | ||
AK098778.1 | - | 12 | 2364 | |||
AK301993.1 | - | 10 | 1478 | |||
BC000367.2 | - | 6 | 924 | |||
BC004333.1 | - | 14 | 2243 | 3630974 | ||
BC010660.1 | - | 9 | 1430 | 3869603 | ||
BC012880.1 | - | 9 | 1358 | 3459009 | ||
BC013614.1 | - | 9 | 1441 | 3880857 | ||
BC015888.2 | - | 9 | 1354 | 3911623 | ||
BC016170.1 | - | 6 | 924 | |||
BC016800.1 | - | 10 | 1382 | 4072487 | ||
BU855133.1 | - | 4 | 631 | |||
BX647566.1 | - | 9 | 1334 | |||
FJ474908.1 | - | 10 | 1404 | |||
M11560.1 | - | 10 | 1404 | |||
M21190.1 | - | 5 | 822 | |||
X05236.1 | - | 9 | 1380 |
Gene Symbol: ALDOAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | M21191.1 | - | LIT2112 ALDOAP2 |
1 | 1463 |