Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163161.1 | NP_001156633.1 | 6 | 822 | ||
NM_010819.4 | NP_034949.3 | 6 | 825 | |||
GenBank mRNA | AF061272.1 | - | 6 | 757 | ||
AK089500.1 | - | 6 | 816 | |||
AK150926.1 | - | 6 | 728 | |||
AK155919.1 | - | 6 | 744 | |||
AK170692.1 | - | 6 | 686 | |||
AK171409.1 | - | 6 | 763 | |||
BC099471.1 | - | 6 | 735 | 30845162 |