Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpeg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010821.1 | NP_034951.1 | 2 | 2332 | ||
GenBank mRNA | AK030216.1 | - | 2 | 2335 | ||
BC049256.1 | - | 1 | 639 | |||
BC129843.1 | - | 1 | 2226 | 40095576 | ||
BC129844.1 | - | 1 | 2226 | 40095579 | ||
L20315.1 | - | 1 | 2234 |