Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010824.2 | NP_034954.2 | 12, 13 | 2240 | ||
GenBank mRNA | AK160229.1 | - | 11, 12 | 1968 | ||
AK220229.1 | - | 11, 12 | 2013 | |||
AY500847.1 | - | 12, 13 | 1994 | |||
BC053912.1 | - | 11, 12 | 1960 | 30044601 | ||
X15313.1 | - | 12, 13 | 2450 |