Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpv17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294322.1 | NP_001281251.1 | 5 | 401 | ||
NM_001294324.1 | NP_001281253.1 | 5 | 430 | |||
NM_008622.6 | NP_032648.1 | 5 | 401 | |||
GenBank mRNA | AK040477.1 | - | 5 | 371 | ||
BC013452.1 | - | 4 | 354 | 4237284 | ||
BY258057.1 | - | 5 | 373 | |||
CK622051.1 | - | 4 | 343 | |||
DV062332.1 | - | 5 | 483 | |||
M36411.1 | - | 5 | 383 |