Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010827.2 | NP_034957.1 | 1 | 352 | ||
GenBank mRNA | AB221639.1 | - | 1 | 134 | ||
AF087035.1 | - | 1 | 352 | |||
AF108216.1 | - | 1 | 134 | |||
AK138917.1 | - | 1 | 379 | |||
BC103592.1 | - | 1 | 259 | 40046034 | ||
BC103593.1 | - | 1 | 259 | 40046035 | ||
BC103594.1 | - | 1 | 255 | 40046036 | ||
BC103623.1 | - | 1 | 259 | 40046489 |