Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010831.2 | NP_034961.2 | 5 | 622 | ||
GenBank mRNA | AK141817.1 | - | 5 | 622 | ||
AK172427.1 | - | 5 | 622 | |||
AK220203.1 | - | 3 | 235 | |||
U11494.2 | - | 5 | 614 |