Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020597.3 | NP_065622.1 | 2 | 115 | ||
GenBank mRNA | U89840.1 | - | 1 | 83 |