Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010833.2 | NP_034963.2 | 6 | 779 | ||
GenBank mRNA | AK088336.1 | - | 6 | 727 | ||
AK133819.1 | - | 6 | 783 | |||
AK145746.1 | - | 6 | 783 | |||
AK150869.1 | - | 6 | 782 | |||
AK153210.1 | - | 6 | 782 | |||
AK157701.1 | - | 6 | 734 | |||
AK216494.1 | - | 1 | 55 | |||
BC003474.1 | - | 6 | 770 | |||
BC047366.1 | - | 6 | 773 | 3711212 | ||
CF584663.1 | - | 267 | ||||
M86390.1 | - | 5 | 559 | |||
S47577.1 | - | 6 | 764 |