Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039934.1 | NP_001035023.1 | 14 | 1667 | ||
NM_008632.2 | NP_032658.2 | 13 | 1574 | |||
GenBank mRNA | AK166380.1 | - | 14 | 1683 | ||
BC048781.1 | - | 9 | 1272 | |||
BC051410.1 | - | 9 | 1266 | |||
BC052446.1 | - | 13 | 1574 | |||
BC150945.1 | - | 13 | 1573 | 9087856 | ||
CN533265.1 | - | 4 | 359 | |||
M21041.1 | - | 14 | 5423 |
Gene Symbol: A730034C02 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042880.1 | - | 8 | 983 |