Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039934.1 | NP_001035023.1 | 11 | 1407 | ||
NM_008632.2 | NP_032658.2 | 11 | 1407 | |||
GenBank mRNA | AK166380.1 | - | 11 | 1423 | ||
BC048781.1 | - | 7 | 1105 | |||
BC051410.1 | - | 7 | 1099 | |||
BC052446.1 | - | 11 | 1407 | |||
BC150945.1 | - | 11 | 1406 | 9087856 | ||
CD348897.1 | - | 4 | 548 | |||
CN533265.1 | - | 1 | 99 | |||
M21041.1 | - | 12 | 5256 |
Gene Symbol: A730034C02 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042880.1 | - | 6 | 816 |