Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038609.2 | NP_001033698.1 | 8, 9 | 978 | ||
NM_001285455.1 | NP_001272384.1 | 5, 6 | 738 | |||
NM_010838.4 | NP_034968.3 | 6, 7 | 804 | |||
GenBank mRNA | AF483518.1 | - | 5, 6 | 619 | ||
AF483519.1 | - | 5, 6 | 619 | |||
AK078167.1 | - | 6, 7 | 776 | |||
AK147636.1 | - | 8, 9 | 939 | |||
BC014748.1 | - | 6, 7 | 742 | 4504850 | ||
M93266.1 | - | 1, 2 | 123 | |||
U12914.1 | - | 8, 9 | 862 | |||
U12915.1 | - | 6, 7 | 688 | |||
U12916.1 | - | 5, 6 | 622 | |||
Z12133.1 | - | 1, 2 | 123 |