Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038609.2 | NP_001033698.1 | 8 | 725 | ||
NM_001285454.1 | NP_001272383.1 | 6 | 551 | |||
NM_001285455.1 | NP_001272384.1 | 5 | 485 | |||
NM_001285456.1 | NP_001272385.1 | 6 | 551 | |||
NM_010838.4 | NP_034968.3 | 6 | 551 | |||
GenBank mRNA | AF483518.1 | - | 5 | 366 | ||
AF483519.1 | - | 5 | 366 | |||
AK078167.1 | - | 6 | 524 | |||
AK147636.1 | - | 8 | 686 | |||
BC014748.1 | - | 6 | 489 | 4504850 | ||
CO432693.1 | - | 6 | 501 | |||
M18775.1 | - | 6 | 552 | |||
M18776.1 | - | 6 | 552 | |||
U12914.1 | - | 8 | 609 | |||
U12915.1 | - | 6 | 435 | |||
U12916.1 | - | 5 | 369 |