Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253877.1 | NP_001240806.1 | 14 | 1956 | ||
NM_001253878.1 | NP_001240807.1 | 14 | 1675 | |||
NM_001253879.1 | NP_001240808.1 | 13 | 1598 | |||
NM_001253880.1 | NP_001240809.1 | 11 | 1313 | |||
NM_013827.3 | NP_038855.2 | 13 | 1594 | |||
GenBank mRNA | AB221622.1 | - | 10 | 996 | ||
AK030752.1 | - | 13 | 1544 | |||
AK031409.1 | - | 15 | 1663 | |||
AK032798.1 | - | 14 | 1917 | |||
AK032819.1 | - | 14 | 1918 | |||
AK137539.1 | - | 15 | 1903 | |||
AK139685.1 | - | 14 | 1676 | |||
BC050826.1 | - | 13 | 1548 | |||
BC092237.1 | - | 13 | 1526 | 30637248 | ||
BC100340.1 | - | 11 | 1244 | 30715034 | ||
S78454.1 | - | 10 | 998 |