Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039657.2 | NP_001034746.1 | 2 | 526 | ||
NM_001039658.2 | NP_001034747.1 | 2 | 526 | |||
NM_001286557.1 | NP_001273486.1 | 3 | 610 | |||
NM_001286558.1 | NP_001273487.1 | 2 | 541 | |||
GenBank mRNA | AB057423.1 | - | 2 | 514 | ||
AF329359.1 | - | 2 | 551 | |||
AF329360.1 | - | 3 | 620 | |||
AK015724.1 | - | 2 | 467 | |||
AK015732.1 | - | 2 | 499 | |||
AK161511.1 | - | 2 | 527 | |||
BC024377.1 | - | 2 | 502 | 4976492 | ||
U67176.1 | - | 4 | 543 | |||
U77383.1 | - | 2 | 163 |