Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164190.1 | NP_001157662.1 | 13 | 1628 | ||
NM_001164191.1 | NP_001157663.1 | 13 | 1519 | |||
NM_001164192.1 | NP_001157664.1 | 12 | 1426 | |||
NM_019926.3 | NP_064310.2 | 13 | 1508 | |||
GenBank mRNA | AF073996.1 | - | 13 | 1509 | ||
AK016445.1 | - | 13 | 1629 | |||
AK048699.1 | - | 1 | 1213 | |||
AK166452.1 | - | 13 | 1487 | |||
AK171475.1 | - | 12 | 1405 | |||
AK220524.1 | - | 3629 | ||||
BC051022.1 | - | 13 | 1477 | |||
BC066847.1 | - | 13 | 1464 | |||
BC090984.1 | - | 13 | 1486 | 30246206 |