Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Laptm4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008640.2 | NP_032666.2 | 4 | 1298 | ||
GenBank mRNA | AK013963.1 | - | 3 | 316 | ||
AK075699.1 | - | 4 | 533 | |||
AK084515.1 | - | 4 | 941 | |||
AK088958.1 | - | 4 | 558 | |||
AK165638.1 | - | 4 | 567 | |||
BC086667.1 | - | 4 | 478 | 30633914 | ||
BC089021.1 | - | 4 | 511 | 6734038 | ||
U34259.1 | - | 5 | 1165 |