Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mast2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042743.2 | NP_001036208.1 | 19 | 2364 | ||
NM_008641.3 | NP_032667.2 | 18 | 2343 | |||
GenBank mRNA | AB093264.1 | - | 9 | 996 | ||
BC060703.1 | - | 19 | 1934 | 6412811 | ||
CK633885.1 | - | 4 | 392 | |||
U02313.1 | - | 18 | 2013 |