Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013604.2 | NP_038632.2 | 3, 4 | 684 | ||
GenBank mRNA | AK086135.1 | - | 3, 4 | 276 | ||
AK159316.1 | - | 2, 3 | 371 | |||
AK163982.1 | - | 3, 4 | 252 | |||
AK165547.1 | - | 3, 4 | 259 | |||
AK165923.1 | - | 3, 4 | 257 | |||
AX775987.1 | - | 3, 4 | 256 | |||
BB662550.1 | - | 3, 4 | 276 | |||
BC023071.1 | - | 3, 4 | 255 | 5355724 | ||
L36962.1 | - | 3, 4 | 254 |