Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bloc1s5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139063.1 | NP_620702.1 | 5 | 581 | ||
GenBank mRNA | AF426433.1 | - | 5 | 581 | ||
AK075808.1 | - | 5 | 527 | |||
AK147762.1 | - | 561 | ||||
AK170614.1 | - | 5 | 531 | |||
BC023184.1 | - | 5 | 600 | 4945742 | ||
BC024720.1 | - | 5 | 538 | 5361934 |