Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013605.2 | NP_038633.1 | 4 | 1585 | ||
GenBank mRNA | BC005441.1 | - | 4 | 1556 | 3601782 | |
BG079301.2 | - | 3 | 214 | |||
M84683.1 | - | 4 | 1535 |