Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prol1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008644.2 | NP_032670.2 | 3 | 195 | ||
GenBank mRNA | AF247816.1 | - | 3 | 197 | ||
BC021158.1 | - | 3 | 199 | |||
BC021401.1 | - | 3 | 197 | 4166946 | ||
BC042902.1 | - | 3 | 193 | 4924369 | ||
U37531.1 | - | 3 | 195 |