Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144514.2 | NP_653097.2 | 2 | 254 | ||
GenBank mRNA | AB076722.1 | - | 3 | 931 | ||
AK082909.1 | - | 2 | 231 | |||
AK133507.1 | - | 2 | 230 | |||
AK147766.1 | - | 92 | ||||
BC030052.1 | - | 2 | 154 | |||
BC038641.1 | - | 2 | 201 | |||
BC051210.1 | - | 2 | 215 | |||
BC132083.1 | - | 2 | 243 | 40130360 | ||
BC132085.1 | - | 2 | 244 | 40130362 | ||
BY337798.1 | - | 2 | 255 | |||
D85430.1 | - | 2 | 224 |