Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mut | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008650.3 | NP_032676.2 | 5, 6 | 1220 | ||
GenBank mRNA | AK146309.1 | - | 5, 6 | 1180 | ||
BC019175.1 | - | 5, 6 | 1202 | 5007694 | ||
X51941.1 | - | 4, 5 | 1131 |