Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mxi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008542.2 | NP_001008542.2 | 6 | 1042 | ||
NM_001008543.2 | NP_001008543.1 | 6 | 729 | |||
NM_010847.3 | NP_034977.1 | 6 | 882 | |||
GenBank mRNA | AK140465.1 | - | 6 | 832 | ||
AK144219.1 | - | 6 | 924 | |||
AK154538.1 | - | 6 | 1032 | |||
BC064453.1 | - | 6 | 1027 | 6810025 | ||
D31824.1 | - | 6 | 882 | |||
L38821.1 | - | 6 | 735 | |||
L38822.1 | - | 6 | 754 | |||
U24673.1 | - | 6 | 715 | |||
U24674.1 | - | 6 | 664 |