Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198914.1 | NP_001185843.1 | 13 | 2131 | ||
NM_010848.3 | NP_034978.3 | 12 | 1774 | |||
GenBank mRNA | AK132250.1 | - | 12 | 1639 | ||
AK137794.1 | - | 6 | 719 | |||
AK137965.1 | - | 12 | 1640 | |||
M12848.1 | - | 12 | 1772 |